Anti-PEA15 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX81197
89301-486EA 623.13 USD
89301-486
Anti-PEA15 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to PEA-15 (C-term)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100FACS: Use at 1:10-50Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: PEA15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR