Anti-KLF16 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX80982
89301-066EA 623.13 USD
89301-066
Anti-KLF16 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to KLF16 (Internal)

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: KLF16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR