Anti-GCG Goat Polyclonal Antibody

Supplier: Genetex
GTX42674
89300-116EA 623.9 USD
89300-116
Anti-GCG Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Glucagon (N-term)

Type: Primary
Antigen: GCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR