Anti-c-myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 289-19510(9E10-F5)]

Supplier: Genetex
GTX44310
89299-896EA 623.11 USD
89299-896
Anti-c-myc (v-myc myelocytomatosis viral oncogene homolog (avian)) Mouse Monoclonal Antibody [clone: 289-19510(9E10-F5)]
Antibodies
Mouse Monoclonal antibody [289-19510(9E10-F5)] to c-myc

Type: Primary
Antigen: c-myc (v-myc myelocytomatosis viral oncogene homolog (avian))
Clonality: Monoclonal
Clone: 289-19510(9E10-F5)
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR