Anti-CD3 Mouse Monoclonal Antibody [clone: MEM-57]

Supplier: Genetex
GTX79934
89298-776EA 513.78 USD
89298-776
Anti-CD3 Mouse Monoclonal Antibody [clone: MEM-57]
Antibodies
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: MEM-57
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR