Anti-CD203c (ectonucleotide pyrophosphatase/phosphodiesterase 3) Mouse Monoclonal Antibody [clone: NP4D6]

Supplier: Genetex
GTX79908
89298-752EA 620.97 USD
89298-752
Anti-CD203c (ectonucleotide pyrophosphatase/phosphodiesterase 3) Mouse Monoclonal Antibody [clone: NP4D6]
Antibodies
PE-conjugated Mouse Monoclonal antibody [NP4D6] to CD203c

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: CD203c (ectonucleotide pyrophosphatase/phosphodiesterase 3)
Clonality: Monoclonal
Clone: NP4D6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR