Anti-CENTA2 Goat Polyclonal Antibody

Supplier: Genetex
GTX89646
89297-912EA 589.79 USD
89297-912
Anti-CENTA2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CENTA2

Preliminary experiments in Human Brain and Heart lysates gave no specific signal but low background (at antibody concentration up to 1µg/ml). We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? In paraffin embedded Human Placenta shows trong staining of cytoplasm in syncytiotrophoblasts.. Recommended concentration, 5-10µg/ml.

Type: Primary
Antigen: CENTA2
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR