Anti-CENTB2 Goat Polyclonal Antibody

Supplier: Genetex
GTX89639
89297-900EA 620.03 USD
89297-900
Anti-CENTB2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CENTB2 / ACAP2

Approx 90kDa band observed in Human T cell line MOLT4 lysates (calculated MW of 88.1kDa according to NP_036419). Recommended concentration: 1-3µg/ml. In paraffin embedded Human Lung shows strong staining of Type II pneumocytes. Recommended concentration, 5-10µg/ml.

Type: Primary
Antigen: CENTB2
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR