Anti-CILP Goat Polyclonal Antibody

Supplier: Genetex
GTX89301
89297-466EA 620.03 USD
89297-466
Anti-CILP Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CILP

Approx 150kDa band observed in rat spinal cord lysates (calculated MW of 133kDa according to human NP_003604.2 and of 132kDa according to Rat NP_001101631.1). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: CILP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR