Anti-PRPF31 Goat Polyclonal Antibody

Supplier: Genetex
GTX89187
89297-310EA 620.03 USD
89297-310
Anti-PRPF31 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PRPF31

Approx 60kDa band observed in lysates of cell lines A431 and HeLa (calculated MW of 55.4kDa according to NP_056444.2). Recommended concentration: 1-3µg/ml. An additional band of unknown identity was also consistently observed at 35kDa. This band was successfully blocked by incubation with the immunising peptide. Also a band of 28kDa was consistently observed, however this band was not blocked by the immunizing peptide and it is therefore a non-specific signal. We call for caution when used for other assays than Western blot.

Type: Primary
Antigen: PRPF31
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR