Anti-DGAT1 Goat Polyclonal Antibody

Supplier: Genetex
GTX89164
89297-278EA 620.03 USD
89297-278
Anti-DGAT1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to DGAT1

Approx 50kDa band observed in lysates of human duodenum (calculated MW of 55.3kDa according to NP_036211.1). Recommended concentration: 0.5-1µg/ml.

Type: Primary
Antigen: DGAT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR