Anti-FADS1 Goat Polyclonal Antibody

Supplier: Genetex
GTX89153
89297-260EA 589.47 USD
89297-260
Anti-FADS1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to FADS1

Approx 50kDa band observed in Human Liver and Lung lysates (calculated MW of 51.9kDa according to NP_037534.2). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: FADS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR