Anti-GALR2 Goat Polyclonal Antibody

Supplier: Genetex
GTX89151
89297-256EA 620.03 USD
89297-256
Anti-GALR2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to GALR2

Approx 40kDa band observed in Human Heart lysates (calculated MW of 41.7kDa according to NP_003848.1). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: GALR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR