Anti-LXN Goat Polyclonal Antibody

Supplier: Genetex
GTX89143
89297-242EA 620.03 USD
89297-242
Anti-LXN Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Latexin

Approx 30kDa band observed in lysates of Human Brain (Cerebellum), Mouse Brain and Rat Brain (calculated MW of 25.8kDa according to NP_064554.2). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: LXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR