Anti-CRP2 / CSRP2 (cysteine and glycine-rich Protein 2) Goat Polyclonal Antibody

Supplier: Genetex
GTX88912
89296-946EA 589.47 USD
89296-946
Anti-CRP2 / CSRP2 (cysteine and glycine-rich Protein 2) Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CRP2 / CSRP2

Approx 26kDa band observed in Human Heart and Kidney lysates and in Mouse and Rat Kidney lysates (calculated MW of 21.0kDa according to NP_001312.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Huber et al, J Biol Chem. 2000 Feb 25;275(8):5504-11; PMID: 10681529). In transfected HEK293 transiently expressing CRP2 a band of approx. 26kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: CRP2 / CSRP2 (cysteine and glycine-rich Protein 2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR