Anti-ASF1A Goat Polyclonal Antibody

Supplier: Genetex
GTX88909
89296-942EA 620.03 USD
89296-942
Anti-ASF1A Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to ASF1A

Approx 21kDa band observed in lysates of cell line Jurkat (calculated MW of 23.0kDa according to NP_054753.1). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: ASF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR