Anti-MARCH6 Goat Polyclonal Antibody

Supplier: Genetex
GTX88852
89296-880EA 589.47 USD
89296-880
Anti-MARCH6 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to TEB4 / MARCH-VI (aa 701 to 712)

Approx 100kDa band observed in Human Lung lysates (calculated MW of 103kDa according to NP_005876.2). Recommended concentration: 0.2-0.5µg/ml. An additional band of unknown identity was also consistently observed at 18kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any such results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?

Type: Primary
Antigen: MARCH6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR