Anti-EXO1 Goat Polyclonal Antibody

Supplier: Genetex
GTX88837
89296-860EA 620.03 USD
89296-860
Anti-EXO1 Goat Polyclonal Antibody
Antibodies
Goat Polyclonal antibody to Exonuclease 1

Approx 90kDa band observed in lysates in cell lines HEK293 and A549 (calculated MW of 89.1kDa according to NP_003677.3). Recommended concentration: 0.2-0.5µg/ml. An additional band of unknown identity was also consistently observed at 38kDa. This band was successfully blocked by incubation with the immunising peptide. We would appreciate any feedback from people in the field - have any such results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? This antibody is expected to recognise all three reported isoforms (NP_003677.3; NP_006018.3 and NP_569082.1)

Type: Primary
Antigen: EXO1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR