Anti-DACH2 Goat Polyclonal Antibody

Supplier: Genetex
GTX88789
89296-806EA 620.03 USD
89296-806
Anti-DACH2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Dachshund homolog 2 / DACH2

Approx 65-70kDa band observed in lysates of cell line NIH3T3 (calculated MW of 65.3kDa according to human NP_444511.1 and of 68.6kDa according to mouse NP_291083.1). Recommended concentration: 0.03-0.1µg/ml.

Type: Primary
Antigen: DACH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR