Anti-RASSF6 Goat Polyclonal Antibody

Supplier: Genetex
GTX88787
89296-804EA 620.03 USD
89296-804
Anti-RASSF6 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to RASSF6

Approx 45kDa band observed in lysates of cell line HepG2 (calculated MW of 43.4kDa according to NP_958834.1). Recommended concentration: 1-3µg/ml. This antibody is expected to recognise both reported isoforms (NP_803876.1; NP_958834.1).

Type: Primary
Antigen: RASSF6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR