Anti-GCKR Goat Polyclonal Antibody

Supplier: Genetex
GTX88432
89296-384EA 620.03 USD
89296-384
Anti-GCKR Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to GCKR

Approx 70kDa band observed in Mouse and Rat Liver lysates (calculated MW of 68.7kDa according to Human NP_001477.1 and of 68.9kDa according to Rat NP_037252.1). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: GCKR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR