Anti-GJA1 Goat Polyclonal Antibody

Supplier: Genetex
GTX88424
89296-376EA 620.03 USD
89296-376
Anti-GJA1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Connexin 43 / GJA1

Approx 40kDa band observed in Rat Brain lysates (calculated MW of 43.0kDa according to Human NP_000156.1 and to Rat NP_036699.1). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: GJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR