Anti-LIG1 Goat Polyclonal Antibody

Supplier: Genetex
GTX88393
89296-326EA 620.03 USD
89296-326
Anti-LIG1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to DNA ligase I

Approx 125kDa band observed in nuclear lysates of cell line HeLa (calculated MW of 102kDa according to NP_000225.1). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: LIG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR