Anti-CLN2 / TPP1 (tripeptidyl peptidase I) Goat Polyclonal Antibody

Supplier: Genetex
GTX88372
89296-286EA 620.03 USD
89296-286
Anti-CLN2 / TPP1 (tripeptidyl peptidase I) Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CLN2 / TPP1

Approx 65kDa band observed in Human Placenta lysates (calculated MW of 61.2kDa according to NP_000382.3). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: CLN2 / TPP1 (tripeptidyl peptidase I)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR