Anti-SLC4A11 Goat Polyclonal Antibody

Supplier: Genetex
GTX88270
89296-194EA 589.27 USD
89296-194
Anti-SLC4A11 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SLC4A11 / BTR1

Approx 90kDa band observed in Human, Mouse and Rat Kidney lysates (calculated MW of 99.6kDa according to NP_114423.1). Recommended concentration: 0.3-1µg/ml. An additional band of unknown identity was also consistently observed at 26kDa. This band was successfully blocked by incubation with the immunizing peptide. We would appreciate any feedback from people in the field - have any such results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?

Type: Primary
Antigen: SLC4A11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR