Anti-MON1A Goat Polyclonal Antibody

Supplier: Genetex
GTX88138
89296-064EA 589.47 USD
89296-064
Anti-MON1A Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to MON1A (aa478-489)

Approx 50kDa band observed in Human Lung, Pancreas and Uterus lysates (calculated MW of 55.5kDa according to NP_001135973.1). Recommended concentration: 1-3µg/ml. This antibody is expected to recognize isoforms a and b (NP_115731.2; NP_001135973.1). Amino acid numbering in name refers to sequence NP_115731.2.

Type: Primary
Antigen: MON1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: 478-489 aa
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR