Anti-GABRG2 Goat Polyclonal Antibody

Supplier: Genetex
GTX88118
89296-040EA 620.03 USD
89296-040
Anti-GABRG2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to GABRG2 (aa400-410)

Approx 50kDa band observed in Mouse and Rat Brain lysates (calculated MW of 55.1kDa according to Mouse NP_032099.1 and 54.1kDa according to Rat NP_899156.1). Recommended concentration: 1-3µg/ml. This antibody is expected to recognize all reported isoforms: NP_944494.1; NP_000807.2; NP_944493.2. Note the aa numbers used in the name are from NP_944494.1.

Type: Primary
Antigen: GABRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR