Anti-CST8 Goat Polyclonal Antibody

Supplier: Genetex
GTX88034
89295-924EA 620.03 USD
89295-924
Anti-CST8 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Cystatin 8 (aa 100-112)

In transfected HEK293 transiently expressing Human CST8 (MYC and DYKDDDDK tagged), a band of approx. 23kDa is observed. No bands are observed in mock-transfected HEK293 and the same band is observed using anti-myc tag antibody. Recommended concentration, 1-3µg/ml.

Type: Primary
Antigen: CST8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR