Anti-SLC2A4 Goat Polyclonal Antibody

Supplier: Genetex
GTX88031
89295-918EA 620.03 USD
89295-918
Anti-SLC2A4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SLC2A4 / GLUT4

Approx 45kDa band observed in Mouse and Rat Heart lysates (calculated MW of 54.8kDa according to Human NP_001033.1 and Mouse NP_033230.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Fischer et al, J Biol Chem. 1997 Mar 14;272(11):7085-92.; PMID: 9054401). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: SLC2A4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR