Anti-SNAP23 Goat Polyclonal Antibody

Supplier: Genetex
GTX88021
89295-906EA 620.03 USD
89295-906
Anti-SNAP23 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SNAP23

Approx 26kDa band observed in Human Placenta lysates (calculated MW of 23.4kDa according to NP_003816.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.1-0.3µg/ml. This antibody is expected to recognize both reported isoforms (NP_003816.2; NP_570710.1).

Type: Primary
Antigen: SNAP23
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR