Anti-NFATC4 Goat Polyclonal Antibody

Supplier: Genetex
GTX89854
89294-272EA 629.87 USD
89294-272
Anti-NFATC4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to NFATC4

No signal obtained yet but low background observed in MOLT-4, Jurkat, A431, HeLa, Human Brain and Human Heart lysates at upto 2µg/ml.

Type: Primary
Antigen: NFATC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR