Anti-NLRP3 Goat Polyclonal Antibody

Supplier: Genetex
GTX89303
89294-052EA 594.69 USD
89294-052
Anti-NLRP3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to CIAS1 / Cryopyrin (Internal)

Preliminary experiments gave an approx 70kDa band in human lymph node and tonsil lysates after 0.3µg/ml antibody staining. Please note that currently we cannot find an explanation in the literature for the band we observe given the calculated size of 118kDa according to NP_004886.3 and 106kDa according to NP_899632.1. The 70kDa band was successfully blocked by incubation with the immunizing peptide. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported? This antibody is expected to recognise both isoform a (NP_004886.3 and NP_001073289.1) and isoform b (NP_899632.1).

Type: Primary
Antigen: NLRP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR