Anti-RNF213 Goat Polyclonal Antibody

Supplier: Genetex
GTX89208
89294-014EA 634.52 USD
89294-014
Anti-RNF213 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to RNF213 / C17orf27 (internal)

Not yet tested - our routinely used western blotting protocol does not allow detection of proteins as large as the calculated size of 374kDa according to NP_065965.3. Therefore we cannot recommend an optimal concentration and the product is investigative grade. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? Have any further splice variants/modified forms been reported?

Type: Primary
Antigen: RNF213
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR