Anti-THRB Goat Polyclonal Antibody

Supplier: Genetex
GTX48511
89293-162EA 594.69 USD
89293-162
Anti-THRB Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to Thrb (zebrafish)

Not yet tested. At this stage we are dependent on researchers in the field for further characterization of this product. Therefore we cannot recommend an optimal concentration and the product is investigative grade. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates? This antibody is not expected to cross-react to Thra.

Type: Primary
Antigen: THRB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR