Anti-HSP104 Rabbit Polyclonal Antibody [clone: 42583]

Supplier: Genetex
GTX79159
89292-274EA 629.01 USD
89292-274
Anti-HSP104 Rabbit Polyclonal Antibody [clone: 42583]
Antibodies
Rabbit Polyclonal antibody to Heat Shock Protein 104

Recommended Starting Dilutions: IHC: 1:100 - 1:200 WB: 1:1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HSP104
Clonality: Polyclonal
Clone: 42583
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR