Anti-IL5 Rat Monoclonal Antibody [clone: 39D10]

Supplier: Genetex
GTX79486
89292-242EA 628.83 USD
89292-242
Anti-IL5 Rat Monoclonal Antibody [clone: 39D10]
Antibodies
Rat monoclonal antibody [39D10] to IL-5

Type: Primary
Antigen: IL5
Clonality: Monoclonal
Clone: 39D10
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR