Anti-FOXD3 Mouse Monoclonal Antibody [clone: 5G9]

Supplier: Genetex
GTX80414
89291-702EA 630.81 USD
89291-702
Anti-FOXD3 Mouse Monoclonal Antibody [clone: 5G9]
Antibodies
Mouse Monoclonal antibody [5G9] to FOXD3

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: FOXD3
Clonality: Monoclonal
Clone: 5G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR