Anti-SLC22A1 Mouse Monoclonal Antibody [clone: 2C5]

Supplier: Genetex
GTX80400
89291-678EA 638.47 USD
89291-678
Anti-SLC22A1 Mouse Monoclonal Antibody [clone: 2C5]
Antibodies
Mouse Monoclonal antibody [2C5] to SLC22A1

Recommended Starting Dilutions:ELISA: 1/10000.WB: 1/500 - 1/2000.FCM: 1/200 - 1/400.Not yet tested in other applications.Determining optimal working dilutions by titration test.

Type: Primary
Antigen: SLC22A1
Clonality: Monoclonal
Clone: 2C5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR