Anti-GPR157 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87984
89291-498EA 629.01 USD
89291-498
Anti-GPR157 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GPR157

We recommend the following starting dilutions: WB: 1:500~1:1000 IF: 1:100~1:500 ELISA: 1:5000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GPR157
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR