Anti-OR10J6 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87892
89291-316EA 629.01 USD
89291-316
Anti-OR10J6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OR10J6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR