Anti-TAS2R14 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87673
89290-890EA 628.83 USD
89290-890
Anti-TAS2R14 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to TAS2R14

  • We recommend the following starting dilutions: WB: 1:500~1:1000 IF: 1:100~1:500 ELISA: 1:10000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TAS2R14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR