Anti-MRPL13 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87614
89290-774EA 629.01 USD
89290-774
Anti-MRPL13 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MRPL13

We recommend the following starting dilutions: WB: 1:500~1:1000 IHC: 1:50~1:100 ELISA: 1:40000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MRPL13
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR