Anti-CACNA1H Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87612
89290-770EA 629.01 USD
89290-770
Anti-CACNA1H Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cav3.2

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:10000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CACNA1H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR