Anti-Amyloid beta (A4) Precursor Protein-binding Family A member 3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87526
89290-606EA 629.01 USD
89290-606
Anti-Amyloid beta (A4) Precursor Protein-binding Family A member 3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Mint3

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:40000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR