Anti-OR6A2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87521
89290-596EA 629.01 USD
89290-596
Anti-OR6A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to OR6A2

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: OR6A2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR