Anti-hnRNP Q (synaptotagmin binding cytoplasmic RNA interacting Protein) Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87520
89290-594EA 629.01 USD
89290-594
Anti-hnRNP Q (synaptotagmin binding cytoplasmic RNA interacting Protein) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to hnRNP Q

We recommend the following starting dilutions: WB: 1:500~1:1000 IF: 1:100~1:500 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: hnRNP Q (synaptotagmin binding cytoplasmic RNA interacting Protein)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR