Anti-STRADA Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87312
89290-184EA 629.01 USD
89290-184
Anti-STRADA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to STRAD

We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:10000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: STRADA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR