Anti-OR5D3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87288
89290-136EA 629.01 USD
89290-136
Anti-OR5D3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: OR5D3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR