Anti-TECR Rabbit Polyclonal Antibody

Supplier: Genetex
GTX87026
89289-620EA 629.01 USD
89289-620
Anti-TECR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GPSN2

We recommend the following starting dilutions: IHC: 1:50~1:100 ELISA: 1:40000Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: TECR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR