Anti-G3BP1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX47441
89288-938EA 653.49 USD
89288-938
Anti-G3BP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to G3BP (N-terminal)

Recommended Dilutions: For Western Blot: Use at 2.5 ug/ml.

Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.


Type: Primary
Antigen: G3BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR